Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 g/mol | C6H7D5ClN3O3 | 25 MG
SDP

Catalog No. 5000303417
Encompass_Preferred

L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of L-histidine (five deuterium atoms) in its hydrochloride hydrate salt form. It is supplied as a white to off-white solid and is used primarily as an isotopic internal standard and tracer for analytical methods, especially mass spectrometry.

  • Deuterium-labeled (d5) for isotope tracing.
  • High purity (≈99.6%).
  • Molecular weight 214.66 g/mol.
  • Chemical formula C6H7D5ClN3O3.
  • Hydrochloride hydrate salt form; stable solid for handling.
  • Suitable as an internal standard and tracer in quantitative mass spectrometry.

Catalog No. 50-003-03417 Supplier Medchemexpress LLC Supplier No. 25MGHYW014423S4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of L-histidine (five deuterium atoms) in its hydrochloride hydrate salt form. It is supplied as a white to off-white solid and is used primarily as an isotopic internal standard and tracer for analytical methods, especially mass spectrometry.

  • Deuterium-labeled (d5) for isotope tracing.
  • High purity (≈99.6%).
  • Molecular weight 214.66 g/mol.
  • Chemical formula C6H7D5ClN3O3.
  • Hydrochloride hydrate salt form; stable solid for handling.
  • Suitable as an internal standard and tracer in quantitative mass spectrometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.