Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine, free base, ≥99%, MP Biomedicals™

Catalog No. ICN10195425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10195425 25 g
ICN10195480 100 g
ICN10195490 500 g
ICN10195491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10195425 Supplier MP Biomedicals, Inc Supplier No. 0210195425
Add to Cart
Add to Cart

Precursor of histamine by action of histidine decarboxylase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 25 g
Formula Weight 155.16
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
EINECSNumber 200-745-3
Recommended Storage Store at Room Temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.