Learn More
Research Products International Corp L-Histidine Hydrochloride Monohydrate, 1 Kilogram
SDP
Catalog No. 50488717
Encompass_Preferred
L-Histidine Hydrochloride Monohydrate is a compound commonly used in life science research, particularly in biochemistry and molecular biology. It is the hydrochloride salt form of L-histidine, an essential amino acid that plays crucial roles in protein synthesis, enzyme catalysis, and neurotransmitter regulation. The monohydrate aspect indicates that it contains one molecule of water per molecule of the compound.
In research, L-Histidine Hydrochloride Monohydrate is often utilized in cell culture media to support cell growth and viability, as well as in protein purification and crystallization processes. 
Its availability in a stable, crystalline form makes it valuable for experimental reproducibility and accuracy in various biological assays and investigations.
Catalog No. 50-488-717 Supplier Research Products International Corp Supplier No. H750801000.0
L-Histidine Hydrochloride Monohydrate is a compound commonly used in life science research, particularly in biochemistry and molecular biology. It is the hydrochloride salt form of L-histidine, an essential amino acid that plays crucial roles in protein synthesis, enzyme catalysis, and neurotransmitter regulation. The monohydrate aspect indicates that it contains one molecule of water per molecule of the compound.
In research, L-Histidine Hydrochloride Monohydrate is often utilized in cell culture media to support cell growth and viability, as well as in protein purification and crystallization processes. 
Its availability in a stable, crystalline form makes it valuable for experimental reproducibility and accuracy in various biological assays and investigations.