Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine Hydrochloride Monohydrate (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP383100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP383-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP383-100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
L-Histidine Hydrochloride is suitable for use in tissue culture systems or as an amino acid standard.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 3.0 to 5.0
Color White
Chloride 18.22 to 18.68
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Loss on Drying 0.2% max.
Other Amino Acids Not Detected
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Specific Rotation 8.9 to 9.5° (+ or -)
State of Solution (%T) 98.0% min.
Alpha Vector Histidine Hydrochloride Monohydrate
Quantity 100 g
Formula Weight 209.64
Percent Purity 99.0 to 101.0%
Physical Form Solid
Chemical Name or Material L-Histidine Hydrochloride Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.