Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Histidine monohydrochloride monohydrate | 5934-29-2 | MFCD00151027 | 25g
Catalog No. NC1224341
Encompass
L-Histidine monohydrochloride monohydrate | Purity: 98.5-101.0% | MW:209.63 | 5934-29-2 | MFCD00151027 | 25g
Catalog No. NC1224341 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H565925G
L-Histidine monohydrochloride monohydrate | Purity: 98.5-101.0% | MW:209.63 | 5934-29-2 | MFCD00151027 | 25g