Learn More
Abcam L-Histidine monohydrochloride monohydrate, Histamine precursor, 25G
Catalog No. 502704816
Encompass_Preferred
MW 209.63 g/mol. Monohydrochloride salt of the amino acid L-histidine. Histamine precursor via histidine decarboxylase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4816 Supplier Abcam Supplier No. AB14623125G
MW 209.63 g/mol. Monohydrochloride salt of the amino acid L-histidine. Histamine precursor via histidine decarboxylase.
The product is subject to the following: Abcam Restricted Use Statement