Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine, MP Biomedicals™

Catalog No. MP210195410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210195410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210195410 Supplier MP Biomedicals, Inc Supplier No. 0210195410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Functions as a precursor of histamine by action of histidine decarboxylase. L-Histidine, MP Biomedicals is an essential amino acid with a basic side chain that contains an imidazole group. It is hydrophilic in character. It is generally used in media prep

Histidine is biosynthesized from ATP, 5-phosphoribosyl-1-pyrophosphate (PRPP), and glutamine. In turn, histidine is degraded to glutamate by histidase, urocanase, and imidazolonepropionase via the formation of urocanate, 4-imidazolone 5-propionate, and N-formiminoglutamate.

  • Has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis
  • Has been shown to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus
  • Utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Solubility Information Soluble 50mg/mL in 0.5 M HCl.
Quantity 10 g
Formula Weight 155.2
Percent Purity ≥99%
Grade Biochemical
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.