Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-histidinol dihydrochloride | 1596-64-1 | MFCD00078058 | >=98.0% | 214.10 g/mol | C6H13Cl2N3O | 1 ML
SDP

Catalog No. 5000302102
Encompass_Preferred

L-histidinol dihydrochloride is the dihydrochloride salt of L-histidinol and is used in research to probe protein synthesis, cell proliferation, and related metabolic pathways. Manufacturer datasheets provide guidance for stock preparation, solubility, formulation options, and storage for research applications.

  • CAS number: 1596-64-1.
  • Molecular formula: C6H13Cl2N3O.
  • Molecular weight: 214.10 g/mol.
  • Soluble in DMSO and water at 100 mg/mL; ultrasonic assistance may be required.
  • Preparation guidance provided for 10 mM solutions and small-volume stocks.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month), sealed.
  • Biological activity: histidyl-tRNA synthetase inhibitor that modulates protein synthesis and cell proliferation.

Catalog No. 50-003-02102 Supplier Medchemexpress LLC Supplier No. HYW01423310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-histidinol dihydrochloride is the dihydrochloride salt of L-histidinol and is used in research to probe protein synthesis, cell proliferation, and related metabolic pathways. Manufacturer datasheets provide guidance for stock preparation, solubility, formulation options, and storage for research applications.

  • CAS number: 1596-64-1.
  • Molecular formula: C6H13Cl2N3O.
  • Molecular weight: 214.10 g/mol.
  • Soluble in DMSO and water at 100 mg/mL; ultrasonic assistance may be required.
  • Preparation guidance provided for 10 mM solutions and small-volume stocks.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month), sealed.
  • Biological activity: histidyl-tRNA synthetase inhibitor that modulates protein synthesis and cell proliferation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.