Learn More
Selleck Chemical LLC L-Homoarginine hydrochloride S5431-25mg
SDP
Catalog No. 503109507
Encompass_Preferred
L-Homoarginine is a specific inhibitor of liver-type alkaline phosphatase
Catalog No. 50-310-9507 Supplier Selleck Chemical LLC Supplier No. S543125MG
L-Homoarginine is a specific inhibitor of liver-type alkaline phosphatase