Learn More
Medchemexpress LLC HY-W018004 100mg Medchemexpress, L-Homocitrulline CAS:1190-49-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW018004100MG
Encompass_Preferred
Medchemexpress, HY-W018004 100mg L-Homocitrulline CAS:1190-49-4 L-Homocitrulline is metabolized to homoarginine through homoargininosuccinate via the urea cycle pathway and its metabolic abnormality could lead to Lysinuric Protein Intolerance (LPI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4638