Learn More
Medchemexpress LLC L-homocitrulline | 1190-49-4 | MFCD00038143 | 99.5% | 189.21 g/mol | C7H15N3O3 | 1 ML
SDP
Catalog No. 5000301600
Encompass_Preferred
L-homocitrulline is an endogenous amino acid metabolite involved in the urea cycle and relevant to studies of homoarginine formation and lysinuric protein intolerance. It is supplied for research use in either aqueous solution (10 mM, 1 mL) or as a solid, with high reported purity and defined storage recommendations for long-term stability.
  • Endogenous metabolite relevant to urea cycle and amino acid metabolism.
  • Available as 10 mM aqueous solution or solid for experimental flexibility.
  • High purity appropriate for analytical and biochemical assays.
  • Defined storage conditions support stability during long-term studies.
  • Suitable for metabolic pathway and biomarker research applications.
Catalog No. 50-003-01600 Supplier Medchemexpress LLC Supplier No. HYW01800410MM/1ML
L-homocitrulline is an endogenous amino acid metabolite involved in the urea cycle and relevant to studies of homoarginine formation and lysinuric protein intolerance. It is supplied for research use in either aqueous solution (10 mM, 1 mL) or as a solid, with high reported purity and defined storage recommendations for long-term stability.
  • Endogenous metabolite relevant to urea cycle and amino acid metabolism.
  • Available as 10 mM aqueous solution or solid for experimental flexibility.
  • High purity appropriate for analytical and biochemical assays.
  • Defined storage conditions support stability during long-term studies.
  • Suitable for metabolic pathway and biomarker research applications.