Learn More
Cayman Chemical L-Homocysteine, Purity: ≥95%, CAS: 6027-13-0, 10 mg
SDP
Catalog No. 502988922
Encompass_Preferred
A thiol-containing amino acid; elevated levels lead to HHcy, which is associated with an increased risk of cardiovascular disease and stroke
Catalog No. 50-298-8922 Supplier Cayman Chemical Supplier No. 3085210 MG
A thiol-containing amino acid; elevated levels lead to HHcy, which is associated with an increased risk of cardiovascular disease and stroke