Learn More
Medchemexpress LLC L-Homocysteine | 6027-13-0 | 98.0% | 135.19 | 25 MG
SDP
Catalog No. 5000454019
Encompass_Preferred
L-Homocysteine is an amino acid with an L configuration and an essential intermediate in normal mammalian metabolism of methionine. It induces upregulation of Cathepsin V, which mediates vascular endothelial inflammation in hyperhomocysteinaemia. It is for research use only and not sold to patients.
  • Amino acid
  • Essential intermediate in normal mammalian metabolism of methionine
  • Induces upregulation of Cathepsin V
  • Can be used in animal modeling to construct a model of vascular endothelial inflammation
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: ketones, aldehydes, acids
  • Initial source: endogenous metabolite human gut microbiota metabolites, microorganisms
  • Upregulation of Cathepsin V mediates vascular endothelial inflammation in hyperhomocysteinaemia
Catalog No. 50-004-54019 Supplier Medchemexpress LLC Supplier No. 25MGHYW010347
L-Homocysteine is an amino acid with an L configuration and an essential intermediate in normal mammalian metabolism of methionine. It induces upregulation of Cathepsin V, which mediates vascular endothelial inflammation in hyperhomocysteinaemia. It is for research use only and not sold to patients.
  • Amino acid
  • Essential intermediate in normal mammalian metabolism of methionine
  • Induces upregulation of Cathepsin V
  • Can be used in animal modeling to construct a model of vascular endothelial inflammation
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: ketones, aldehydes, acids
  • Initial source: endogenous metabolite human gut microbiota metabolites, microorganisms
  • Upregulation of Cathepsin V mediates vascular endothelial inflammation in hyperhomocysteinaemia