Learn More
Medchemexpress LLC L-Homocysteine | 6027-13-0 | 98.0% | 135.19 | 50 MG
SDP
Catalog No. 5000367061
Encompass_Preferred
L-Homocysteine is an amino acid with an L configuration, and an essential intermediate in the normal mammalian metabolism of methionine. It induces the upregulation of Cathepsin V, which mediates vascular endothelial inflammation in hyperhomocysteinaemia. This product is for research use only.
  • Essential intermediate in methionine metabolism
  • Induces upregulation of Cathepsin V
  • Suitable for animal modeling to construct vascular endothelial inflammation models
Catalog No. 50-003-67061 Supplier Medchemexpress LLC Supplier No. HYW01034750MG
L-Homocysteine is an amino acid with an L configuration, and an essential intermediate in the normal mammalian metabolism of methionine. It induces the upregulation of Cathepsin V, which mediates vascular endothelial inflammation in hyperhomocysteinaemia. This product is for research use only.
  • Essential intermediate in methionine metabolism
  • Induces upregulation of Cathepsin V
  • Suitable for animal modeling to construct vascular endothelial inflammation models