Learn More
Medchemexpress LLC L-homocysteine-d4 | 331665-12-4 | MFCD01317494 | 94.9% | 139.21 | C4H5D4NO2S | 1 MG
SDP
Catalog No. 5000284716
Encompass_Preferred
L-Homocysteine-d4 is the deuterium-labeled form of L-homocysteine provided as a neat solid for use as an internal standard and tracer in quantitative analytical methods. It is intended for research and analytical applications such as LC-MS, GC-MS, and NMR, enabling isotope-dilution quantitation and method calibration.
  • Deuterium-labeled L-homocysteine for isotope-dilution quantitation.
  • High purity suitable for analytical standards and reference materials.
  • Supplied as a white to off-white solid stable under sealed storage at 4°C.
  • Soluble in water and DMSO with ultrasonic assistance for sample preparation.
  • Known chemical formula and molecular weight facilitate accurate mass confirmation during analysis.
Catalog No. 50-002-84716 Supplier Medchemexpress LLC Supplier No. HYW6537331MG
Add to Cart
Add to Cart
L-Homocysteine-d4 is the deuterium-labeled form of L-homocysteine provided as a neat solid for use as an internal standard and tracer in quantitative analytical methods. It is intended for research and analytical applications such as LC-MS, GC-MS, and NMR, enabling isotope-dilution quantitation and method calibration.
  • Deuterium-labeled L-homocysteine for isotope-dilution quantitation.
  • High purity suitable for analytical standards and reference materials.
  • Supplied as a white to off-white solid stable under sealed storage at 4°C.
  • Soluble in water and DMSO with ultrasonic assistance for sample preparation.
  • Known chemical formula and molecular weight facilitate accurate mass confirmation during analysis.