Learn More
Medchemexpress LLC L-homocysteine, s-[2-[(1-iminoethyl)amino]ethyl]-, phosphate (1:1) | 438542-15-5 | 95.9% | 10 MG
SDP
Catalog No. 5000232491
Encompass_Preferred
GW274150 phosphate is the phosphate salt of GW274150, a selective, orally active, NADPH-dependent inhibitor of inducible nitric oxide synthase (iNOS) provided for research use. It is supplied as solid or as a 10 mM solution and is characterized for activity against human and rodent iNOS.
  • Selective iNOS inhibitor (human iNOS IC50 = 2.19 μM; Kd = 40 nM).
  • Available as phosphate salt in solid and solution formats.
  • Pack sizes include 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • High purity by HPLC (95.9%).
  • Molecular weight 317.30 g/mol.
  • Suitable for in vitro and in vivo iNOS inhibition studies.
Catalog No. 50-002-32491 Supplier Medchemexpress LLC Supplier No. HY12119A10MG
GW274150 phosphate is the phosphate salt of GW274150, a selective, orally active, NADPH-dependent inhibitor of inducible nitric oxide synthase (iNOS) provided for research use. It is supplied as solid or as a 10 mM solution and is characterized for activity against human and rodent iNOS.
  • Selective iNOS inhibitor (human iNOS IC50 = 2.19 μM; Kd = 40 nM).
  • Available as phosphate salt in solid and solution formats.
  • Pack sizes include 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • High purity by HPLC (95.9%).
  • Molecular weight 317.30 g/mol.
  • Suitable for in vitro and in vivo iNOS inhibition studies.