Learn More
Medchemexpress LLC HY-101404A 10mg Medchemexpress, L-Homocysteine thiolactone (hydrochloride) CAS:31828-68-9 Purity:>98%
SDP
Catalog No. 501932111
Encompass_Preferred
Medchemexpress, HY-101404A 10mg L-Homocysteine thiolactone (hydrochloride) CAS:31828-68-9 L-Homocysteine thiolactone hydrochloride is an intramolecular thioester of homocysteine; prevents translational incorporation of homocysteine into proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2111 Supplier Medchemexpress LLC Supplier No. HY101404A10MG
Medchemexpress, HY-101404A 10mg L-Homocysteine thiolactone (hydrochloride) CAS:31828-68-9 L-Homocysteine thiolactone hydrochloride is an intramolecular thioester of homocysteine; prevents translational incorporation of homocysteine into proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.