Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-homocysteine thiolactone hydrochloride | 31828-68-9 | MFCD00065494 | 100.0% | 153.63 g·mol⁻¹ | C4H8ClNOS | 25 MG
SDP

Catalog No. 5000249121
Encompass_Preferred

L-Homocysteine thiolactone hydrochloride is the intramolecular thioester of homocysteine supplied as a research reagent. It functions as an amine-reactive compound used to probe homocysteine metabolism and protein post-translational modification in biochemical and cellular studies.

  • Acts as an amine-reactive agent that can modify lysine residues.
  • Used to study homocysteine metabolism and protein incorporation.
  • Molecular formula: C4H8ClNOS; molecular weight: 153.63 g·mol⁻¹.
  • CAS number: 31828-68-9.
  • High stated purity (≈99.99%).
  • Available in small pack sizes such as 25 mg for laboratory use.

Catalog No. 50-002-49121 Supplier Medchemexpress LLC Supplier No. HY101404A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Homocysteine thiolactone hydrochloride is the intramolecular thioester of homocysteine supplied as a research reagent. It functions as an amine-reactive compound used to probe homocysteine metabolism and protein post-translational modification in biochemical and cellular studies.

  • Acts as an amine-reactive agent that can modify lysine residues.
  • Used to study homocysteine metabolism and protein incorporation.
  • Molecular formula: C4H8ClNOS; molecular weight: 153.63 g·mol⁻¹.
  • CAS number: 31828-68-9.
  • High stated purity (≈99.99%).
  • Available in small pack sizes such as 25 mg for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.