Learn More
Medchemexpress LLC HY-W011690 100mg Medchemexpress, L-Homocystine CAS:626-72-2 Purity:>98%
SDP
Catalog No. 501934554
Encompass_Preferred
Medchemexpress, HY-W011690 100mg L-Homocystine CAS:626-72-2 L-Homocystine is the oxidized member of the L-homocysteine. Homocysteine is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor and endoplasmatic reticulum-stress inducer used to study cardiovascular disease mechanisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4554 Supplier Medchemexpress LLC Supplier No. HYW011690100MG
Medchemexpress, HY-W011690 100mg L-Homocystine CAS:626-72-2 L-Homocystine is the oxidized member of the L-homocysteine. Homocysteine is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor and endoplasmatic reticulum-stress inducer used to study cardiovascular disease mechanisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.