Learn More
Medchemexpress LLC L-HOMOCYSTINE-D8 | 182755-41-5 | 99.9% | 276.40 | 1 MG
SDP
Catalog No. 5000452575
Encompass_Preferred
L-Homocystine-d8 is the deuterium labeled L-Homocystine. L-Homocystine is the oxidized member of L-homocysteine, which is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor, and endoplasmatic reticulum-stress inducer. It is used to study cardiovascular disease mechanisms.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52575 Supplier Medchemexpress LLC Supplier No. HYW011690S1MG
L-Homocystine-d8 is the deuterium labeled L-Homocystine. L-Homocystine is the oxidized member of L-homocysteine, which is a pro-thrombotic factor, vasodilation impairing agent, pro-inflammatory factor, and endoplasmatic reticulum-stress inducer. It is used to study cardiovascular disease mechanisms.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.