Learn More
Apexbio Technology LLC L-Homopropargyl Glycine(Synonyms: HPG, Homopropargylglycine, L-HPG, L-Homopropargylglycine, L-2-Amino-5-hexynoic acid), 50mg, CAS: 98891-36-2.
SDP
Catalog No. 5000061044
Encompass_Preferred
L-Homopropargyl Glycine (CAS 98891-36-2) abbreviated as Hpg is a noncanonical amino acid structurally analogous to methionine featuring an alkyne functional group Upon incorporation into newly synthesized proteins during translation Hpg enables selective labeling of nascent proteins through copper-catalyzed alkyne-azide cycloaddition ( click chemistry ) This fluorescent tagging strategy permits temporal tracking of protein synthesis dynamics in mammalian cells via pulse-chase labeling Such a methodology facilitates direct imaging of protein translation activities including those relevant to neuronal function and synaptic plasticity aiding the investigation of compartment-specific protein synthesis
Catalog No. 50-000-61044 Supplier Apexbio Technology LLC Supplier No. C340650
L-Homopropargyl Glycine (CAS 98891-36-2) abbreviated as Hpg is a noncanonical amino acid structurally analogous to methionine featuring an alkyne functional group Upon incorporation into newly synthesized proteins during translation Hpg enables selective labeling of nascent proteins through copper-catalyzed alkyne-azide cycloaddition ( click chemistry ) This fluorescent tagging strategy permits temporal tracking of protein synthesis dynamics in mammalian cells via pulse-chase labeling Such a methodology facilitates direct imaging of protein translation activities including those relevant to neuronal function and synaptic plasticity aiding the investigation of compartment-specific protein synthesis