Learn More
Medchemexpress LLC L-homopropargylglycine (2S)-2-amino-5-hexynoic acid | 98891-36-2 | MFCD11052896 | 98.0% | 127.14 g/mol | C6H9NO2 | 5 MG
SDP
Catalog No. 5000222756
Encompass_Preferred
L-homopropargylglycine is an alkyne-bearing amino acid analog used as a PROTAC linker and for bioorthogonal click chemistry. The compound contains a terminal alkyne for azide-alkyne cycloaddition and is supplied as a small, crystalline reagent suitable for synthetic and conjugation workflows.
  • Alkyne-functional amino acid ideal for click chemistry applications.
  • Suitable as a PROTAC linker or for probe conjugation.
  • High purity of 98.0% supports reliable synthesis.
  • Low molecular weight (127.14 g/mol) simplifies handling and analysis.
  • Stable when stored sealed at recommended cold temperatures.
  • Available in small pack sizes for research-scale use.
Catalog No. 50-002-22756 Supplier Medchemexpress LLC Supplier No. HY1403455MG
L-homopropargylglycine is an alkyne-bearing amino acid analog used as a PROTAC linker and for bioorthogonal click chemistry. The compound contains a terminal alkyne for azide-alkyne cycloaddition and is supplied as a small, crystalline reagent suitable for synthetic and conjugation workflows.
  • Alkyne-functional amino acid ideal for click chemistry applications.
  • Suitable as a PROTAC linker or for probe conjugation.
  • High purity of 98.0% supports reliable synthesis.
  • Low molecular weight (127.14 g/mol) simplifies handling and analysis.
  • Stable when stored sealed at recommended cold temperatures.
  • Available in small pack sizes for research-scale use.