Learn More
Medchemexpress LLC HY-W002292 100mg Medchemexpress, L-Homoserine CAS:672-15-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW002292100MG
Encompass_Preferred
Medchemexpress, HY-W002292 100mg L-Homoserine CAS:672-15-1 L-Homoserine is a non - protein amino acid, which is an important biosynthetic intermediate of threonine, methionine and lysine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4472