Learn More
AdipoGen L-Homoserine lactone hydrobromide (5 g)
SDP
Supplier:  AdipoGen CDXH0310G005
Encompass_Preferred
Chemical. CAS: 15295-77-9. Formula: C4H7NO2 . HBr. MW: 182.02. L-Homoserine lactone hydrobromide is used as a building block. It can be used to prepare beta-ketoamide N-acylated-L-homoserine lactones (AHLs) as quorum sensing molecules, p-coumaroyl-HSL (homoserine lactone), calpain and lipid peroxidation inhibitors and pseudopeptide inhibitors for Ras farnesyl-protein transferase, as well as selenomethionine.
Catalog No. 50-205-4502