Learn More
AdipoGen L-Homoserine lactone HCl
SDP
Catalog No. 502560695
Encompass_Preferred
Chemical. CAS 2185-03-7. Formula C4H7NO2 . HCl. MW 137.56. L-Homoserine lactone is a synthetic building block or intermediate used in the synthesis of organoselenium chemistry intermediates and analogs of bacterial quorum-sensing signaling molecules.
Catalog No. 50-256-0695 Supplier AdipoGen Supplier No. CDXH0281G005
Chemical. CAS 2185-03-7. Formula C4H7NO2 . HCl. MW 137.56. L-Homoserine lactone is a synthetic building block or intermediate used in the synthesis of organoselenium chemistry intermediates and analogs of bacterial quorum-sensing signaling molecules.