Learn More
Cayman Chemical L-hydroxy Arginine (acetate), Purity: ≥95%, CAS: 53598-01-9, 10 mg
SDP
Catalog No. 502894093
Encompass_Preferred
A substrate for NOS in the formation of NO; as been used as a biomarker for reduced NO formation in patients with cardiovascular disease and metabolic syndrome
Catalog No. 50-289-4093 Supplier Cayman Chemical Supplier No. 8030010 MG
A substrate for NOS in the formation of NO; as been used as a biomarker for reduced NO formation in patients with cardiovascular disease and metabolic syndrome