Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Hydroxy arginine d 100mg | 133565-48-7 | 100 MG
SDP

Catalog No. 5000308139
Encompass_Preferred

L-hydroxy arginine (dihydrochloride) is an intermediate in the catabolism of L-arginine and a substrate for nitric oxide synthase. Supplied as a powdered dihydrochloride salt for laboratory research, it is intended for research use only.

  • CAS number: 133565-48-7.
  • Molecular weight: 263.12 g/mol.
  • Chemical formula: C6H16Cl2N4O3.
  • Physical form: Powder.
  • Salt form: Dihydrochloride.
  • Storage: Powder -80°C for 2 years or -20°C for 1 year; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended use: For research use only.

Catalog No. 50-003-08139 Supplier Medchemexpress LLC Supplier No. HY115411A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-hydroxy arginine (dihydrochloride) is an intermediate in the catabolism of L-arginine and a substrate for nitric oxide synthase. Supplied as a powdered dihydrochloride salt for laboratory research, it is intended for research use only.

  • CAS number: 133565-48-7.
  • Molecular weight: 263.12 g/mol.
  • Chemical formula: C6H16Cl2N4O3.
  • Physical form: Powder.
  • Salt form: Dihydrochloride.
  • Storage: Powder -80°C for 2 years or -20°C for 1 year; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended use: For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.