Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-α-Hydroxyglutaric acid-13C5 disodium salt, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111003831 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3831 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3831 Supplier MilliporeSigma Supelco Supplier No. 8055910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Linear Formula 13C5H6 Na2O5
Synonym -2-Hydroxypentanedioic acid-1,2,3,4,5-13C5 disodium salt; Disodium -2-hydroxyglutarate-13C5
Type Analytical Standard
Quantity 10 mg
Formula Weight 197.04
Percent Purity ≥98% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.