Learn More
Medchemexpress LLC L-Hydroxyproline-d3 | 1356016-86-8 | 99.9% | 134.15 | 5 MG
SDP
Catalog No. 5000403785
Encompass_Preferred
L-Hydroxyproline-d3 is the deuterium labeled L-Hydroxyproline. L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-03785 Supplier Medchemexpress LLC Supplier No. HY40135S5MG
L-Hydroxyproline-d3 is the deuterium labeled L-Hydroxyproline. L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.