Learn More
Medchemexpress LLC L-Hydroxyproline (Standard) | 51-35-4 | 99.9% | 131.13 | 50 MG
SDP
Catalog No. 5000401430
Encompass_Preferred
L-Hydroxyproline (Standard) is the analytical standard of L-Hydroxyproline. This product is intended for research and analytical applications. L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Structure classification: Alkaloids, Pyrrole Alkaloids.
  • Initial source: Endogenous metabolite, Disease markers, Endocrine diseases, Nervous System Disorder, Immune System Disorder.
Catalog No. 50-004-01430 Supplier Medchemexpress LLC Supplier No. HY40135R50MG
L-Hydroxyproline (Standard) is the analytical standard of L-Hydroxyproline. This product is intended for research and analytical applications. L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Structure classification: Alkaloids, Pyrrole Alkaloids.
  • Initial source: Endogenous metabolite, Disease markers, Endocrine diseases, Nervous System Disorder, Immune System Disorder.