Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Hyocyamine, free base, 100%, MP Biomedicals™

Catalog No. ICN15131601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15131601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15131601 Supplier MP Biomedicals, Inc Supplier No. 0215131601

Chemical Identifiers

CAS 101-31-5
Molecular Formula C17H23NO3
Molecular Weight (g/mol) 289.38
MDL Number MFCD00067306,MFCD00022622,MFCD00022622
InChI Key RKUNBYITZUJHSG-VFSICIBPSA-N
Synonym s-atropinium, duboisine, l-tropine tropate, s-atropinium cation, s-atropinium 1+, 3-endo-8-methyl-8-azoniabicyclo 3.2.1 oct-3-yl 2s-3-hydroxy-2-phenylpropanoate, 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl 2s-3-hydroxy-2-phenylpropanoate, 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl 2s-3-oxidanyl-2-phenyl-propanoate, 2s-3-hydroxy-2-phenylpropanoic acid 1r,5s-8-methyl-8-azoniabicyclo 3.2.1 octan-3-yl ester
PubChem CID 638022
IUPAC Name (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate
SMILES CN1[C@H]2CC[C@@H]1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1

Specifications

Color White
Assay Percent Range 1
Quantity 1 g
Formula Weight 289.37
Physical Form Crystalline Powder
Chemical Name or Material L-Hyocyamine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.