Learn More
Medchemexpress LLC L-Hyoscyamine sulfa 25mg
SDP
Catalog No. 5000669446
Encompass_Preferred
L-Hyoscyamine sulfate (Daturine sulfate) a natural plant tropane alkaloid is a potent and competitive muscarinic receptor (MR) antagonist L-Hyoscyamine sulfate is a levo-isomer to Atropine (HY-B1205)[1][2]
Catalog No. 50-006-69446 Supplier Medchemexpress LLC Supplier No. 25MGHYN0471A
L-Hyoscyamine sulfate (Daturine sulfate) a natural plant tropane alkaloid is a potent and competitive muscarinic receptor (MR) antagonist L-Hyoscyamine sulfate is a levo-isomer to Atropine (HY-B1205)[1][2]