Learn More
Medchemexpress LLC L-Hyoscyamine sulfate | 620-61-1 | 338.41 | 20 MG
SDP
Catalog No. 5000388776
Encompass_Preferred
L-Hyoscyamine sulfate (Daturine sulfate) is a natural plant tropane alkaloid. It functions as a potent and competitive muscarinic receptor (MR) antagonist. This compound is also recognized as a levo-isomer to Atropine.
  • Potent and competitive muscarinic receptor (MR) antagonist
  • Natural plant tropane alkaloid
  • Levo-isomer to atropine
Catalog No. 50-003-88776 Supplier Medchemexpress LLC Supplier No. HYN0471A20MG
L-Hyoscyamine sulfate (Daturine sulfate) is a natural plant tropane alkaloid. It functions as a potent and competitive muscarinic receptor (MR) antagonist. This compound is also recognized as a levo-isomer to Atropine.
  • Potent and competitive muscarinic receptor (MR) antagonist
  • Natural plant tropane alkaloid
  • Levo-isomer to atropine