Learn More
Medchemexpress LLC L-hyoscyamine sulfate | 620-61-1 | MFCD00078244 | 98.3% | 338.41 g·mol⁻¹ | C17H23NO3·1/2H2SO4 | 10 MG
SDP
Catalog No. 5000295822
Encompass_Preferred
L-hyoscyamine sulfate is the levo-isomer of atropine, a tropane alkaloid that acts as a competitive muscarinic receptor antagonist. Supplied as an analytical standard for research and analytical applications; not for human use.
  • Analytical standard for research and analytical applications.
  • Levo-isomer of atropine with muscarinic receptor antagonist activity.
  • Reported purity 98.3% (SKU-specific value listed on product page).
  • CAS number 620-61-1 and molecular weight 338.41 g·mol⁻¹.
  • Provided as a 10 mg quantity suitable for analytical assays and method development.
Catalog No. 50-002-95822 Supplier Medchemexpress LLC Supplier No. HYN0471A10MG
L-hyoscyamine sulfate is the levo-isomer of atropine, a tropane alkaloid that acts as a competitive muscarinic receptor antagonist. Supplied as an analytical standard for research and analytical applications; not for human use.
  • Analytical standard for research and analytical applications.
  • Levo-isomer of atropine with muscarinic receptor antagonist activity.
  • Reported purity 98.3% (SKU-specific value listed on product page).
  • CAS number 620-61-1 and molecular weight 338.41 g·mol⁻¹.
  • Provided as a 10 mg quantity suitable for analytical assays and method development.