Learn More
Medchemexpress LLC L-Hyoscyamine sulfate (Daturine sulfate) | 620-61-1 | 338.41 | 5 MG
SDP
Catalog No. 5000389685
Encompass_Preferred
L-Hyoscyamine sulfate (Daturine sulfate), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. It is a levo-isomer to Atropine. It is for research use only and not sold to patients.
  • Potent and competitive muscarinic receptor (MR) antagonist.
  • Natural plant tropane alkaloid.
  • Levo-isomer to Atropine.
Catalog No. 50-003-89685 Supplier Medchemexpress LLC Supplier No. HYN0471A5MG
L-Hyoscyamine sulfate (Daturine sulfate), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. It is a levo-isomer to Atropine. It is for research use only and not sold to patients.
  • Potent and competitive muscarinic receptor (MR) antagonist.
  • Natural plant tropane alkaloid.
  • Levo-isomer to Atropine.