Learn More
Medchemexpress LLC L-Inosine | 21138-24-9 | 99.9% | 268.23 | 100 MG
SDP
Catalog No. 5000363644
Encompass_Preferred
L-Inosine is the L-configuration of Inosine, an endogenous purine nucleoside produced by the catabolism of adenosine.
  • L-configuration of Inosine
  • Endogenous purine nucleoside
  • Possesses anti-inflammatory effects
  • Exhibits antinociceptive properties
  • Acts as an immunomodulator
  • Provides neuroprotective benefits
Catalog No. 50-003-63644 Supplier Medchemexpress LLC Supplier No. HY148166100MG
L-Inosine is the L-configuration of Inosine, an endogenous purine nucleoside produced by the catabolism of adenosine.
  • L-configuration of Inosine
  • Endogenous purine nucleoside
  • Possesses anti-inflammatory effects
  • Exhibits antinociceptive properties
  • Acts as an immunomodulator
  • Provides neuroprotective benefits