Learn More
Medchemexpress LLC L-Inosine | 21138-24-9 | 99.9% | 268.23 | 25 MG
SDP
Catalog No. 5000363750
Encompass_Preferred
L-Inosine is the L-configuration of Inosine, an endogenous purine nucleoside produced by the catabolism of adenosine. It exhibits anti-inflammatory, antinociceptive, immunomodulatory, and neuroprotective effects.
  • Exhibits anti-inflammatory effects
  • Exhibits antinociceptive effects
  • Exhibits immunomodulatory effects
  • Exhibits neuroprotective effects
  • L-configuration of inosine
  • Endogenous purine nucleoside
  • Produced by the catabolism of adenosine
Catalog No. 50-003-63750 Supplier Medchemexpress LLC Supplier No. 25MGHY148166
L-Inosine is the L-configuration of Inosine, an endogenous purine nucleoside produced by the catabolism of adenosine. It exhibits anti-inflammatory, antinociceptive, immunomodulatory, and neuroprotective effects.
  • Exhibits anti-inflammatory effects
  • Exhibits antinociceptive effects
  • Exhibits immunomodulatory effects
  • Exhibits neuroprotective effects
  • L-configuration of inosine
  • Endogenous purine nucleoside
  • Produced by the catabolism of adenosine