Learn More
Medchemexpress LLC L-inosine | 21138-24-9 | MFCD00673221 | 99.9% | 268.23 g/mol | C10H12N4O5 | 10 MG
SDP
Catalog No. 5000286747
Encompass_Preferred
L-Inosine is the L-configuration of the endogenous purine nucleoside inosine, provided as a high-purity research chemical for biochemical and pharmacological studies. It is commonly used in investigations of anti-inflammatory, immunomodulatory, antinociceptive, and neuroprotective effects, and is supplied with documentation to support identity and purity.
  • High purity: 99.91% (as reported)
  • Suitable for in vitro biochemical and pharmacological research
  • Available in small, accurately dosed research packs (e.g., 10 mg)
  • Provided with certificate of analysis and product datasheet
  • Soluble in DMSO for easy preparation of stock solutions
Catalog No. 50-002-86747 Supplier Medchemexpress LLC Supplier No. HY14816610MG
L-Inosine is the L-configuration of the endogenous purine nucleoside inosine, provided as a high-purity research chemical for biochemical and pharmacological studies. It is commonly used in investigations of anti-inflammatory, immunomodulatory, antinociceptive, and neuroprotective effects, and is supplied with documentation to support identity and purity.
  • High purity: 99.91% (as reported)
  • Suitable for in vitro biochemical and pharmacological research
  • Available in small, accurately dosed research packs (e.g., 10 mg)
  • Provided with certificate of analysis and product datasheet
  • Soluble in DMSO for easy preparation of stock solutions