Learn More
Refrigiwear Inc. 2860R Regular Gold/Blk Large IRON TUFF GLOVE
SDP
Supplier:  Refrigiwear Inc. 2860RGBKLAR
Encompass_Preferred
With 4-times the strength of natural leather, Iron-Tuff® Insulated Leather Gloves can take on even the toughest tasks, no matter how low the temperature drops. With 200g fiberfill plus foam insulation over a soft tricot lining, these thermal work gloves help you give cold the finger, all the way down to -30�F. Kevlar® reinforced seams, thumb crotch & finger tips on these insulated work gloves add durability while the pre-curved ergonomic fit reduces hand fatigue. Warm. Tough. Iconic. The Iron-Tuff® Collection has been proven in the cold for over 65 years. From the Alaska Pipeline to every step of getting food to America's tables, Iron-Tuff® has been there. Extreme durability, legendary warmth - it's the original way to Master the Cold. Comfort Rating: -30�F 200g Fiberfill plus foam insulation with tricot lining Synthetic leather palm is 4 times stronger than natural leather Silicone grip pattern on palm & fingers Kevlar® reinforced seams, thumb crotch & finger ti
Catalog No. NC3404856