Learn More
Medchemexpress LLC L-Isoleucine-13C6,15N | 202468-35-7 | 138.12 | 5 MG
SDP
Catalog No. 5000416395
Encompass_Preferred
L-Isoleucine-13C6,15N is a 13C and 15N labeled L-isoleucine, which is a nonpolar hydrophobic and essential amino acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16395 Supplier Medchemexpress LLC Supplier No. HYN0771S95MG
L-Isoleucine-13C6,15N is a 13C and 15N labeled L-isoleucine, which is a nonpolar hydrophobic and essential amino acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.