Learn More
Medchemexpress LLC L-Isoleucine-15N | 59935-30-7 | 132.17 | 25 MG
SDP
Catalog No. 5000390245
Encompass_Preferred
L-Isoleucine-15N is a 15N-labeled form of L-Isoleucine. L-Isoleucine itself is characterized as a nonpolar, hydrophobic, and essential amino acid. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90245 Supplier Medchemexpress LLC Supplier No. 25MGHYN0771S2
L-Isoleucine-15N is a 15N-labeled form of L-Isoleucine. L-Isoleucine itself is characterized as a nonpolar, hydrophobic, and essential amino acid. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.