Learn More
Medchemexpress LLC L-Isoleucine-15N | 59935-30-7 | 132.17 | 5 MG
SDP
Catalog No. 5000416211
Encompass_Preferred
L-Isoleucine-15N is the 15N-labeled L-Isoleucine, which is a nonpolar hydrophobic and essential amino acid. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for use as tracers during drug development.
  • Used as a tracer for various analyses.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS techniques.
  • Stable heavy isotope-labeled compound.
  • Essential amino acid.
Catalog No. 50-004-16211 Supplier Medchemexpress LLC Supplier No. HYN0771S25MG
L-Isoleucine-15N is the 15N-labeled L-Isoleucine, which is a nonpolar hydrophobic and essential amino acid. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for use as tracers during drug development.
  • Used as a tracer for various analyses.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS techniques.
  • Stable heavy isotope-labeled compound.
  • Essential amino acid.