Learn More
Medchemexpress LLC L-Isoleucine | 73-32-5 | MFCD00064222 | 131.18 | 5 G
SDP
Catalog No. 5000417119
Encompass_Preferred
L-Isoleucine is an orally active branched-chain amino acid, the L-enantiomer of isoleucine. It functions as a metabolite in various organisms, including Saccharomyces cerevisiae, Escherichia coli, plants, humans, algae, and mice. This compound is involved in regulating the inflammatory response, offering protection against pathogens both in vivo and in vitro.
  • Orally active branched-chain amino acid
  • Regulates inflammatory response
  • Offers protection against pathogens
  • High purity (99.82%)
  • White to off-white solid appearance
  • Soluble in water and PBS
Catalog No. 50-004-17119 Supplier Medchemexpress LLC Supplier No. HYN07715G
L-Isoleucine is an orally active branched-chain amino acid, the L-enantiomer of isoleucine. It functions as a metabolite in various organisms, including Saccharomyces cerevisiae, Escherichia coli, plants, humans, algae, and mice. This compound is involved in regulating the inflammatory response, offering protection against pathogens both in vivo and in vitro.
  • Orally active branched-chain amino acid
  • Regulates inflammatory response
  • Offers protection against pathogens
  • High purity (99.82%)
  • White to off-white solid appearance
  • Soluble in water and PBS