Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Isoleucine-d1 | 127290-60-2 | 132.18 | 25 MG
SDP

Catalog No. 5000389160
Encompass_Preferred

L-Isoleucine-d is a deuterium-labeled L-Isoleucine, which is a nonpolar hydrophobic and essential amino acid. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has stable heavy isotopes of hydrogen, carbon, and other elements incorporated.
  • Appearance: Solid, white to off-white.
  • Purity: 99.48%.

Catalog No. 50-003-89160 Supplier Medchemexpress LLC Supplier No. 25MGHYN0771S8
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Isoleucine-d is a deuterium-labeled L-Isoleucine, which is a nonpolar hydrophobic and essential amino acid. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has stable heavy isotopes of hydrogen, carbon, and other elements incorporated.
  • Appearance: Solid, white to off-white.
  • Purity: 99.48%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.