Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Isoleucine-d10 | 35045-71-7 | 141.23 | 5 MG
SDP

Catalog No. 5000416379
Encompass_Preferred

L-Isoleucine-d10 is the deuterium labeled L-Isoleucine. L-isoleucine is a nonpolar hydrophobic amino acid and an essential amino acid. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: H2O : 8.33 mg/mL (58.98 mM; Need ultrasonic)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).

Catalog No. 50-004-16379 Supplier Medchemexpress LLC Supplier No. HYN0771S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Isoleucine-d10 is the deuterium labeled L-Isoleucine. L-isoleucine is a nonpolar hydrophobic amino acid and an essential amino acid. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: H2O : 8.33 mg/mL (58.98 mM; Need ultrasonic)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.