Learn More
Medchemexpress LLC L-isoleucine-d10 | 35045-71-7 | MFCD00144617 | 98.0% | 141.23 | C6H3D10NO2 | 10 MG
SDP
Catalog No. 5000296553
Encompass_Preferred
L-Isoleucine-d10 is the deuterium-labeled form of the essential amino acid l-isoleucine, provided as a stable-isotope reagent for analytical and labeling applications. It is a white to off-white solid with high isotopic enrichment and purity appropriate for mass spectrometry and metabolic tracing.
  • Deuterium-labeled (d10) analog of l-isoleucine.
  • High purity (98.0%) suitable for analytical applications.
  • Soluble in water; may require ultrasonic assistance.
  • White to off-white solid for convenient handling.
  • Suitable as an internal standard for LC-MS/GC-MS and metabolite tracing.
Catalog No. 50-002-96553 Supplier Medchemexpress LLC Supplier No. HYN0771S310MG
L-Isoleucine-d10 is the deuterium-labeled form of the essential amino acid l-isoleucine, provided as a stable-isotope reagent for analytical and labeling applications. It is a white to off-white solid with high isotopic enrichment and purity appropriate for mass spectrometry and metabolic tracing.
  • Deuterium-labeled (d10) analog of l-isoleucine.
  • High purity (98.0%) suitable for analytical applications.
  • Soluble in water; may require ultrasonic assistance.
  • White to off-white solid for convenient handling.
  • Suitable as an internal standard for LC-MS/GC-MS and metabolite tracing.