Learn More
Medchemexpress LLC L-Isoleucine-¹⁵N,d10 | 73-32-5 | 142.23 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0771S75MG
Encompass_Preferred
L-Isoleucine-15N,d10 is an isotopically labeled form of L-Isoleucine, an essential amino acid. This compound is enriched with both deuterium (d10) and nitrogen-15 (15N) isotopes, making it highly valuable for various research applications. It is commonly employed in studies involving protein structure and dynamics, metabolic pathways, and nuclear magnetic resonance (NMR) spectroscopy, where isotopic labeling helps in tracing and identifying specific molecules and their interactions.
  • Isotopically labeled for research applications.
  • Enriched with deuterium and nitrogen-15 isotopes.
  • Useful for tracing metabolic pathways.
  • Ideal for NMR spectroscopy studies.
  • Supports research in protein structure and dynamics.
Catalog No. 50-004-16104