Learn More
Medchemexpress LLC L-Isoleucine (Standard) | 73-32-5 | 131.18 | 25 MG
SDP
Catalog No. 5000389837
Encompass_Preferred
L-Isoleucine (Standard) is the analytical standard of L-Isoleucine, intended for research and analytical applications. It is an orally active branched chain amino acid and the L-enantiomer of isoleucine. Acting as a metabolite in various organisms, it also regulates inflammatory responses to protect against pathogens. This compound serves as a reference standard with a supplied assay.
  • Analytical standard of L-Isoleucine
  • Intended for research and analytical applications
  • Orally active branched chain amino acid
  • Acts as a metabolite in Saccharomyces cerevisiae, Escherichia coli, plants, humans, algae, and mice
  • Regulates inflammatory responses to protect against pathogens
  • Used as a reference standard with a supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-89837 Supplier Medchemexpress LLC Supplier No. 25MGHYN0771R
L-Isoleucine (Standard) is the analytical standard of L-Isoleucine, intended for research and analytical applications. It is an orally active branched chain amino acid and the L-enantiomer of isoleucine. Acting as a metabolite in various organisms, it also regulates inflammatory responses to protect against pathogens. This compound serves as a reference standard with a supplied assay.
  • Analytical standard of L-Isoleucine
  • Intended for research and analytical applications
  • Orally active branched chain amino acid
  • Acts as a metabolite in Saccharomyces cerevisiae, Escherichia coli, plants, humans, algae, and mice
  • Regulates inflammatory responses to protect against pathogens
  • Used as a reference standard with a supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS