Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Isoserine, 98%

Catalog No. AAH6094503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6094503 1 g
AAH60945MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6094503 Supplier Thermo Scientific Chemicals Supplier No. H6094503
Only null left
Add to Cart

CAS: 632-13-3 | C3H7NO3 | 105.09 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-13-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD03428326
InChI Key BMYNFMYTOJXKLE-REOHCLBHSA-N
Synonym l-isoserine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine, s-2-hydroxy-b-alanine
PubChem CID 6999797
SMILES [NH3+]C[C@H](O)C([O-])=O

Specifications

Melting Point 197°C to 198°C
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-azaniumyl-2-hydroxypropanoate
Quantity 1 g
Formula Weight 105.09
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material L-Isoserine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.