Learn More
Enzo Life Sciences L-JNKi1 (1mg)
SDP
Catalog No. 502010740
Encompass_Preferred
This L-stereoisomer is a cell permeable JNK (c-Jun N-terminal kinase) inhibitor. For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter. Neuroprotective agent for stroke. Inhibits the interaction between JNK (JNK-1, -2 and –3) and its substrate with the same IC50 (in vitro IC50 ~ 1µM), but is degraded more readily than the D-stereoisomer, thus requiring higher treatment concentrations in vitro and in vivo. Alternative name: JNK inhibitor 1 (L-stereoisomer), c Jun N terminal kinase peptide inhibitor 1, L stereoisomer. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0740 Supplier Enzo Life Sciences Supplier No. BMLEI3540001
This L-stereoisomer is a cell permeable JNK (c-Jun N-terminal kinase) inhibitor. For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter. Neuroprotective agent for stroke. Inhibits the interaction between JNK (JNK-1, -2 and –3) and its substrate with the same IC50 (in vitro IC50 ~ 1µM), but is degraded more readily than the D-stereoisomer, thus requiring higher treatment concentrations in vitro and in vivo. Alternative name: JNK inhibitor 1 (L-stereoisomer), c Jun N terminal kinase peptide inhibitor 1, L stereoisomer. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.